Anti-CLEC11A Mouse monoclonal antibody unconjugated [clone: MM0533-6D5]
Lieferant: Abcam
ABCAAB90238-50EA 869 EUR
ABCAAB90238-50
Anti-CLEC11A Mouse monoclonal antibody unconjugated [clone: MM0533-6D5]
Antikörper
Type: Primary
Antigen: CLEC11A (C-type lectin domain family 11, member A)
Clonality: monoclonal
Clone: MM0533-6D5
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human