Anti-SELENBP1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB90135-100EA 888 EUR
ABCAAB90135-100
Anti-SELENBP1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SELENBP1 (selenium binding protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR