Anti-IL10RA Mouse Monoclonal Antibody [clone: MM0359-7Q35]

Lieferant: Abcam
ABCAAB89883-100EA 834 EUR
ABCAAB89883-100
Anti-IL10RA Mouse Monoclonal Antibody [clone: MM0359-7Q35]
Antikörper
Type: Primary
Antigen: IL10RA (interleukin 10 receptor, alpha)
Clonality: monoclonal
Clone: MM0359-7Q35
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR