Anti-LILRB1 Mouse polyclonal antibody unconjugated

ABCAAB67532-50EA 672 EUR
ABCAAB67532-50
Anti-LILRB1 Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: LILRB1 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR