Anti-B4GALT3 Mouse Polyclonal Antibody

Lieferant: Abcam
ABCAAB67476-50EA 837 EUR
ABCAAB67476-50
Anti-B4GALT3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: B4GALT3 (UDP-Gal:betaGlcNAc beta 1,4- galactosyltransferase, polypeptide 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR