Anti-DDIT4L Mouse polyclonal antibody unconjugated

ABCAAB67431-50EA 666 EUR
ABCAAB67431-50
Anti-DDIT4L Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: DDIT4L (DNA-damage-inducible transcript 4-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR