Anti-DIRAS2 Mouse polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB67430-50EA 877 EUR
ABCAAB67430-50
Anti-DIRAS2 Mouse polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DIRAS2 (DIRAS family, GTP-binding RAS-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat