Anti-SH2D1B Mouse Polyclonal Antibody

Lieferant: Abcam
ABCAAB67417-50EA 877 EUR
ABCAAB67417-50
Anti-SH2D1B Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SH2D1B (SH2 domain containing 1B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR