Anti-TMC6 Rabbit polyclonal antibody unconjugated

ABCAAB67326-100EA 634 EUR
ABCAAB67326-100
Anti-TMC6 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: TMC6 (transmembrane channel-like 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR