Anti-LGI3 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB67321-100EA 665 EUR
ABCAAB67321-100
Anti-LGI3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LGI3 (leucine-rich repeat LGI family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat