Anti-GRIK1 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB67317-100EA 902 EUR
ABCAAB67317-100
Anti-GRIK1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GRIK1 (glutamate receptor, ionotropic, kainate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR