Anti-SLITRK2 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB67304-100EA 877 EUR
ABCAAB67304-100
Anti-SLITRK2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLITRK2 (SLIT and NTRK-like family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR