Anti-LGI2 Rabbit polyclonal antibody unconjugated

ABCAAB67301-100EA 688 EUR
ABCAAB67301-100
Anti-LGI2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: LGI2 (leucine-rich repeat LGI family, member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR