Anti-LAMP1 Rabbit Polyclonal Antibody (Cy3®)
ABCAAB67283-100EA 672 EUR
ABCAAB67283-100
Anti-LAMP1 Rabbit Polyclonal Antibody (Cy3®)
Type: Primary
Antigen: LAMP1 (lysosomal-associated membrane protein 1)
Clonality: polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen