Anti-UBE2E3 Mouse Polyclonal Antibody

Lieferant: Abcam
ABCAAB66257-50EA 837 EUR
ABCAAB66257-50
Anti-UBE2E3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: UBE2E3 (ubiquitin-conjugating enzyme E2E 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR