Anti-Neurotensin Receptor 1 Guinea pig Polyclonal Antibody
Lieferant: Abcam
ABCAAB66216-100EA 696 EUR
ABCAAB66216-100
Anti-Neurotensin Receptor 1 Guinea pig Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Neurotensin Receptor 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Guinea pig
Isotype: IgG
Reactivity: Rat