Anti-COX7A2L Rabbit polyclonal antibody unconjugated

ABCAAB66107-100EA 634 EUR
ABCAAB66107-100
Anti-COX7A2L Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: COX7A2L (cytochrome c oxidase subunit VIIa polypeptide 2 like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR