Anti-Parainfluenza Virus type 3 Mouse Monoclonal Antibody [clone: M02122321]

Lieferant: Abcam
ABCAAB49756-250EA 706 EUR
ABCAAB49756-250
Anti-Parainfluenza Virus type 3 Mouse Monoclonal Antibody [clone: M02122321]
Antikörper
Type: Primary
Antigen: Parainfluenza Virus type 3
Clonality: monoclonal
Clone: M02122321
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR