Anti-PF4 Mouse Monoclonal Antibody [clone: RTO]

Lieferant: Abcam
ABCAAB49735-100EA 898 EUR
ABCAAB49735-100
Anti-PF4 Mouse Monoclonal Antibody [clone: RTO]
Antikörper
Type: Primary
Antigen: PF4 (platelet factor 4)
Clonality: monoclonal
Clone: RTO
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR