Anti-TFPI Rabbit polyclonal antibody Biotin

ABCAAB49562-150EA 666 EUR
ABCAAB49562-150
Anti-TFPI Rabbit polyclonal antibody Biotin
Type: Primary
Antigen: TFPI (tissue factor pathway inhibitor (lipoprotein-associated coagulation inhibitor))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR