Anti-F2 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Abcam
ABCAAB49552-150EA 877 EUR
ABCAAB49552-150
Anti-F2 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: F2 (coagulation factor II (thrombin))
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR