Anti-IgE Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BE5]
Lieferant: Abcam
ABCAAB49434-100EA 877 EUR
ABCAAB49434-100
Anti-IgE Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BE5]
Antikörper
Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone: BE5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human