Anti-FMRFamide Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB10352-50EA 840 EUR
ABCAAB10352-50
Anti-FMRFamide Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FMRFamide
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR