Anti-Podoplanin / gp36 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB10274-100EA 877 EUR
ABCAAB10274-100
Anti-Podoplanin / gp36 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Podoplanin / gp36
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR