Anti-Delta Opioid Receptor Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB10272-50EA 848 EUR
ABCAAB10272-50
Anti-Delta Opioid Receptor Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Delta Opioid Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR