Anti-P2RX3 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB10269-50EA 860 EUR
ABCAAB10269-50
Anti-P2RX3 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: P2RX3 (purinergic receptor P2X, ligand-gated ion channel, 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR