Anti-DDX5 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB10261-100EA 922 EUR
ABCAAB10261-100
Anti-DDX5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DDX5 (DEAD (Asp-Glu-Ala-Asp) box helicase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR