Anti-P2RX1 Rabbit Polyclonal Antibody

ABCAAB10248-50EA 682 EUR
ABCAAB10248-50
Anti-P2RX1 Rabbit Polyclonal Antibody
Type: Primary
Antigen: P2RX1 (purinergic receptor P2X, ligand-gated ion channel, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR