Anti-TNNI3 Mouse Monoclonal Antibody [clone: 4C2]
Lieferant: Abcam
ABCAAB10231-200EA 926 EUR
ABCAAB10231-200
Anti-TNNI3 Mouse Monoclonal Antibody [clone: 4C2]
Antikörper
Type: Primary
Antigen: TNNI3 (troponin I type 3 (cardiac))
Clonality: monoclonal
Clone: 4C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen