Anti-HRP Mouse Monoclonal Antibody [clone: 2H11]
Lieferant: Abcam
ABCAAB10183-500EA 882 EUR
ABCAAB10183-500
Anti-HRP Mouse Monoclonal Antibody [clone: 2H11]
Antikörper
Type: Primary
Antigen: HRP
Clonality: monoclonal
Clone: 2H11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestellen