Anti-VPS29 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB10160-100EA 882 EUR
ABCAAB10160-100
Anti-VPS29 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPS29 (vacuolar protein sorting 29 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR