Anti-ARHGEF18 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB10152-100EA 877 EUR
ABCAAB10152-100
Anti-ARHGEF18 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARHGEF18 (Rho/Rac guanine nucleotide exchange factor (GEF) 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR