Anti-HIF3A Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB10134-100EA 891 EUR
ABCAAB10134-100
Anti-HIF3A Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HIF3A (hypoxia inducible factor 3, alpha subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR