Anti-FAM9C Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00171484-B01EA 394 EUR
ABNOH00171484-B01
Anti-FAM9C Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FAM9C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR