Anti-SH2D1B Mouse Monoclonal Antibody [clone: 2B4]
Lieferant: Abnova
ABNOH00117157-M04EA 394 EUR
ABNOH00117157-M04
Anti-SH2D1B Mouse Monoclonal Antibody [clone: 2B4]
Antikörper
Type: Primary
Antigen: SH2D1B
Clonality: Monoclonal
Clone: 2B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human