Anti-POMGNT2 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00084892-B01PEA 394 EUR
ABNOH00084892-B01P
Anti-POMGNT2 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POMGNT2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human