Anti-TSSK3 Mouse Monoclonal Antibody [clone: 6H4]

Lieferant: Abnova
ABNOH00081629-M06EA 394 EUR
ABNOH00081629-M06
Anti-TSSK3 Mouse Monoclonal Antibody [clone: 6H4]
Antikörper
Type: Primary
Antigen: TSSK3
Clonality: Monoclonal
Clone: 6H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR