Anti-SPRYD7 Mouse Monoclonal Antibody [clone: 2G4]

Lieferant: Abnova
ABNOH00057213-M01EA 394 EUR
ABNOH00057213-M01
Anti-SPRYD7 Mouse Monoclonal Antibody [clone: 2G4]
Antikörper
Type: Primary
Antigen: SPRYD7
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR