Anti-NTMT1 Mouse Polyclonal Antibody
Lieferant: Abnova
ABNOH00028989-B01PEA 394 EUR
ABNOH00028989-B01P
Anti-NTMT1 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NTMT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human