Anti-PRPF19 Mouse Monoclonal Antibody [clone: 2E5]

Lieferant: Abnova
ABNOH00027339-M07EA 394 EUR
ABNOH00027339-M07
Anti-PRPF19 Mouse Monoclonal Antibody [clone: 2E5]
Antikörper
Type: Primary
Antigen: PRPF19
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR