Anti-B3GNT3 Mouse Monoclonal Antibody [clone: 1A2]

Lieferant: Abnova
ABNOH00010331-M01EA 394 EUR
ABNOH00010331-M01
Anti-B3GNT3 Mouse Monoclonal Antibody [clone: 1A2]
Antikörper
Type: Primary
Antigen: B3GNT3
Clonality: Monoclonal
Clone: 1A2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR