Anti-AKR1A1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00010327-D01PEA 380 EUR
ABNOH00010327-D01P
Anti-AKR1A1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AKR1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR