Anti-TNFRSF6B Mouse Monoclonal Antibody [clone: 1F1]

Lieferant: Abnova
ABNOH00008771-M01EA 394 EUR
ABNOH00008771-M01
Anti-TNFRSF6B Mouse Monoclonal Antibody [clone: 1F1]
Antikörper
Type: Primary
Antigen: TNFRSF6B
Clonality: Monoclonal
Clone: 1F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR