Anti-CAMK1 Mouse Monoclonal Antibody [clone: 2B6]

Lieferant: Abnova
ABNOH00008536-M02EA 394 EUR
ABNOH00008536-M02
Anti-CAMK1 Mouse Monoclonal Antibody [clone: 2B6]
Antikörper
Type: Primary
Antigen: CAMK1
Clonality: Monoclonal
Clone: 2B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR