Anti-COPS3 Mouse Polyclonal Antibody

Lieferant: Abnova
ABNOH00008533-A01EA 258 EUR
ABNOH00008533-A01
Anti-COPS3 Mouse Polyclonal Antibody
Antikörper
Type: Primary
Antigen: COPS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR