Anti-ENC1 Mouse Monoclonal Antibody [clone: 3B1]

Lieferant: Abnova
ABNOH00008507-M02EA 394 EUR
ABNOH00008507-M02
Anti-ENC1 Mouse Monoclonal Antibody [clone: 3B1]
Antikörper
Type: Primary
Antigen: ENC1
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR