Anti-EZR Mouse Monoclonal Antibody [clone: 1E11]

Lieferant: Abnova
ABNOH00007430-M04EA 394 EUR
ABNOH00007430-M04
Anti-EZR Mouse Monoclonal Antibody [clone: 1E11]
Antikörper
Type: Primary
Antigen: EZR
Clonality: Monoclonal
Clone: 1E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR