Anti-SERPINF1 Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00005176-D01PEA 380 EUR
ABNOH00005176-D01P
Anti-SERPINF1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SERPINF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR