Anti-ME1 Mouse Monoclonal Antibody [clone: 3B4]

Lieferant: Abnova
ABNOH00004199-M02EA 394 EUR
ABNOH00004199-M02
Anti-ME1 Mouse Monoclonal Antibody [clone: 3B4]
Antikörper
Type: Primary
Antigen: ME1
Clonality: Monoclonal
Clone: 3B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR