Anti-LPL Rabbit Polyclonal Antibody

Lieferant: Abnova
ABNOH00004023-D01EA 380 EUR
ABNOH00004023-D01
Anti-LPL Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR